Learn More
Medchemexpress LLC 4H-Pyrido[1,2-a]pyrimidin-4-one, 3-[2-[4-(6-fluoro-1,2-benzisoxazol-3-yl)-1-piperidinyl]ethyl-... | 1020719-76-9 | 99.4% | 414.51 | 50 MG
SDP
Catalog No. 5000343128
Encompass_Preferred
Risperidone-d4 is the deuterium-labeled version of Risperidone. Risperidone functions as a serotonin 5-HT2 receptor blocker, a P-Glycoprotein inhibitor, and a potent dopamine D2 receptor antagonist. It exhibits Ki values of 4.8 nM for the 5-HT2A receptor and 5.9 nM for the dopamine D2 receptor. This product is intended for research use only and is not sold to patients.
  • Can be utilized as a tracer.
  • Serves as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes of hydrogen, carbon, and other elements into drug molecules, primarily for quantitation as tracers during drug development.
  • Deuteration has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-43128 Supplier Medchemexpress LLC Supplier No. HY110232500UG
Risperidone-d4 is the deuterium-labeled version of Risperidone. Risperidone functions as a serotonin 5-HT2 receptor blocker, a P-Glycoprotein inhibitor, and a potent dopamine D2 receptor antagonist. It exhibits Ki values of 4.8 nM for the 5-HT2A receptor and 5.9 nM for the dopamine D2 receptor. This product is intended for research use only and is not sold to patients.
  • Can be utilized as a tracer.
  • Serves as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes of hydrogen, carbon, and other elements into drug molecules, primarily for quantitation as tracers during drug development.
  • Deuteration has the potential to influence the pharmacokinetic and metabolic profiles of drugs.