Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4H-Pyrido[1,2-a]pyrimidin-4-one, 3-(2-hydroxyethyl)-2-methyl-9-(phenylmethoxy)- | 1008796-22-2 | 97.5% | C18H18N2O3 | 250 MG
SDP

Catalog No. 5000358426
Encompass_Preferred

Paliperidone impurity 1 is a chemical compound identified as 4H-Pyrido[1,2-a]pyrimidin-4-one, 3-(2-hydroxyethyl)-2-methyl-9-(phenylmethoxy)-, with the molecular formula C18H18N2O3. Supplied as a light yellow to yellow solid, it boasts a purity of 97.5%. This product is strictly for research purposes and has not been fully validated for medical applications.

  • Identified as 4H-Pyrido[1,2-a]pyrimidin-4-one, 3-(2-hydroxyethyl)-2-methyl-9-(phenylmethoxy)-
  • High purity of 97.5%
  • Available as a light yellow to yellow solid
  • Storage conditions for powder: -20°C for 3 years, 4°C for 2 years
  • Storage conditions in solvent: -80°C for 6 months, -20°C for 1 month
  • For research use only

Catalog No. 50-003-58426 Supplier Medchemexpress LLC Supplier No. 250MGHY75265
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Paliperidone impurity 1 is a chemical compound identified as 4H-Pyrido[1,2-a]pyrimidin-4-one, 3-(2-hydroxyethyl)-2-methyl-9-(phenylmethoxy)-, with the molecular formula C18H18N2O3. Supplied as a light yellow to yellow solid, it boasts a purity of 97.5%. This product is strictly for research purposes and has not been fully validated for medical applications.

  • Identified as 4H-Pyrido[1,2-a]pyrimidin-4-one, 3-(2-hydroxyethyl)-2-methyl-9-(phenylmethoxy)-
  • High purity of 97.5%
  • Available as a light yellow to yellow solid
  • Storage conditions for powder: -20°C for 3 years, 4°C for 2 years
  • Storage conditions in solvent: -80°C for 6 months, -20°C for 1 month
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.