Learn More
Enzo Life Sciences Hapten 4-hydroxy-3-nitrophenyl acetyl NP Recombinant monoclonal antibody (B1-8) (Mouse IgG3 lambda) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS5250200
Encompass_Preferred
Clone: B1-8. Isotype: IgG3λ. Immunogen: Hapten-protein conjugated NP12-chicken gammaglobulin (CG). Applications: Flow Cytometry, WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-3051