Learn More
Cayman Chemical 4hdrox3Methoxphenlglc 10mg
SDP
Catalog No. 502984839
Encompass_Preferred
A metabolite of norepinephrine used as a biochemical index of recent sympathetic nervous system activity
Catalog No. 50-298-4839 Supplier Cayman Chemical Supplier No. 1971410 MG
A metabolite of norepinephrine used as a biochemical index of recent sympathetic nervous system activity