Learn More
Cayman Chemical 4hydroxy Diclofenac
SDP
Catalog No. 502890111
Encompass_Preferred
An active metabolite of diclofenac; inhibits COX activity (IC50 = 32 nM) and reduces PGE2 production (IC50 = 17 nM) in isolated human rheumatoid synovial cells
Catalog No. 50-289-0111 Supplier Cayman Chemical Supplier No. 100085185 MG
An active metabolite of diclofenac; inhibits COX activity (IC50 = 32 nM) and reduces PGE2 production (IC50 = 17 nM) in isolated human rheumatoid synovial cells