Learn More
Cayman Chemical 4-hydroxy DiPT (hydrochloride), Purity: ≥95%, CAS: 63065-90-7, 10 mg
SDP
Catalog No. 502951663
Encompass_Preferred
An analytical reference standard categorized as a tryptamine; induces the HTR in mice; substitutes for DOM in a two-lever drug discrimination test in rats; intended for research and forensic applications
Catalog No. 50-295-1663 Supplier Cayman Chemical Supplier No. 1131210 MG
An analytical reference standard categorized as a tryptamine; induces the HTR in mice; substitutes for DOM in a two-lever drug discrimination test in rats; intended for research and forensic applications