Learn More
Cayman Chemical 4-hydroxy Hexenal-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 148706-05-2, 1 mg
SDP
Catalog No. 502889391
Encompass_Preferred
An internal standard for the quantification of 4-hydroxy hexenal by GC- or LC-MS
Catalog No. 50-288-9391 Supplier Cayman Chemical Supplier No. 100076571 MG
An internal standard for the quantification of 4-hydroxy hexenal by GC- or LC-MS