Learn More
Cayman Chemical 4hydroxy Hexenald3
SDP
Catalog No. 502889775
Encompass_Preferred
An internal standard for the quantification of 4-hydroxy hexenal by GC- or LC-MS
Catalog No. 50-288-9775 Supplier Cayman Chemical Supplier No. 10007657100 UG
An internal standard for the quantification of 4-hydroxy hexenal by GC- or LC-MS