Learn More
Cayman Chemical 4hydroxy Hexenald3
SDP
Supplier:  Cayman Chemical 10007657500 UG
Encompass_Preferred
An internal standard for the quantification of 4-hydroxy hexenal by GC- or LC-MS
Catalog No. 50-288-9386