Learn More
Cayman Chemical 4hydroxy Nonal GlutathIon 50UG
SDP
Catalog No. 502894226
Encompass_Preferred
An internal standard for the quantification of 4-HNE-GSH by GC- or LC-MS
Catalog No. 50-289-4226 Supplier Cayman Chemical Supplier No. 900087650 UG
An internal standard for the quantification of 4-HNE-GSH by GC- or LC-MS