Learn More
Cayman Chemical 4-hydroxy Nonenal Glutathione (trifluoroacetate salt), Purity: ≥95%, 5 mg
SDP
Catalog No. 502890456
Encompass_Preferred
A major adduct formed by the reaction of 4-HNE with GSH; 4-HNE-GSH levels in liver, plasma, or isolated cells can serve as biomarkers for oxidative stress
Catalog No. 50-289-0456 Supplier Cayman Chemical Supplier No. 106275 MG
A major adduct formed by the reaction of 4-HNE with GSH; 4-HNE-GSH levels in liver, plasma, or isolated cells can serve as biomarkers for oxidative stress