Learn More
Cayman Chemical 4-hydroxy Tolbutamide, Purity: ≥98%, CAS: 5719-85-7, 10 mg
SDP
Catalog No. 502889857
Encompass_Preferred
A metabolite of tolbutamide; has been used as a marker of CYP2C9 activity in rat hepatocytes
Catalog No. 50-288-9857 Supplier Cayman Chemical Supplier No. 1000852010 MG
A metabolite of tolbutamide; has been used as a marker of CYP2C9 activity in rat hepatocytes