Learn More
Cayman Chemical 4-Hydroxyphenylglyoxylic Acid, Purity: ≥95%, CAS: 15573-67-8, 100 mg
SDP
Catalog No. 502916448
Encompass_Preferred
A CPT1 inhibitor and an active metabolite of oxfenicine; inhibits CPT1 in isolated rat liver mitochondria from 200 to 1,000 µM; inhibits oleate oxidation in isolated rat hepatocytes
Catalog No. 50-291-6448 Supplier Cayman Chemical Supplier No. 38121100 MG
A CPT1 inhibitor and an active metabolite of oxfenicine; inhibits CPT1 in isolated rat liver mitochondria from 200 to 1,000 µM; inhibits oleate oxidation in isolated rat hepatocytes