Learn More
Cayman Chemical 4Hydroyphenylglyoylc ACD 10mg
SDP
Catalog No. 502915908
Encompass_Preferred
A CPT1 inhibitor and an active metabolite of oxfenicine; inhibits CPT1 in isolated rat liver mitochondria from 200 to 1,000 µM; inhibits oleate oxidation in isolated rat hepatocytes
Catalog No. 50-291-5908 Supplier Cayman Chemical Supplier No. 3812110 MG
A CPT1 inhibitor and an active metabolite of oxfenicine; inhibits CPT1 in isolated rat liver mitochondria from 200 to 1,000 µM; inhibits oleate oxidation in isolated rat hepatocytes