Learn More
Electron Microscopy Sciences Silicon Chip Specimen Supports
SDP
Catalog No. NC2602217
Encompass
Silicon Chip Specimen Supports has a siper smoth polished. Has surface properties equal to glass, are opaque, and posess low electircal resistance. Useful for applications where the surfaces of carbon or aluminum stubs may cause interference. With a surface roughness less than 1 nm, resistance of 1-30 Ohms and a wafer thickness of 460-530 pm.
Catalog No. NC2602217 Supplier Electron Microscopy Sciences Supplier No. 7189309
Silicon Chip Specimen Supports has a siper smoth polished. Has surface properties equal to glass, are opaque, and posess low electircal resistance. Useful for applications where the surfaces of carbon or aluminum stubs may cause interference. With a surface roughness less than 1 nm, resistance of 1-30 Ohms and a wafer thickness of 460-530 pm.