Learn More
Cole-Parmer Solid-design 4 in cable tie, white 1000/bag
Supplier: Cole-Parmer 0683052
These ties are injection-molded of 6/6 nylon, so they wont corrode or stretch. They have a durable locking mechanism that holds securely, even when used to bundle vibrating or flexing materials. These ties continue to hold tension even in oily spray or wet environments. Ties easily can be applied by hand or using a tensioning tool. Tools can be purchased separately.