Learn More
Ace Glass, Inc. O-ring, CAPFE, fits 100mm grooved flange
SDP
Catalog No. 509558094
Encompass_Preferred
CAPFE o-ring with 100mm grooved flange, is an ACE-exclusive o-ring with a resilient rubber core encased in a thick, non-porous FEP/PTFE. CAPFE has excellent compression set and chemical compatibility, and low gas and water permeability.
Catalog No. 50-955-8094 Supplier Ace Glass, Inc. Supplier No. 7855880
CAPFE o-ring with 100mm grooved flange, is an ACE-exclusive o-ring with a resilient rubber core encased in a thick, non-porous FEP/PTFE. CAPFE has excellent compression set and chemical compatibility, and low gas and water permeability.