Learn More
California Pacific Lab & Consulting 4IN ECO FUNNEL
SDP
Catalog No. NC9287530
Encompass_Preferred
Eco funnel, 4 in. dia. w/53B screw cap, Made of HDPE and has an easy use lid, gasket, positive latch, filter, and an 11 in. stem to effectively reduce chemical evaporation of hazardous solvents in the lab, 250mL
Catalog No. NC9287530 Supplier California Pacific Lab & Consulting Supplier No. EF453B
Eco funnel, 4 in. dia. w/53B screw cap, Made of HDPE and has an easy use lid, gasket, positive latch, filter, and an 11 in. stem to effectively reduce chemical evaporation of hazardous solvents in the lab, 250mL