Learn More
California Pacific Lab & Consulting 4IN ECO FUNNEL 38-400 RED
SDP
Catalog No. NC3451535
Encompass_Preferred
4" ECO Funnel® with 38-400 cap adapter for 38mm plastic jugsSize:4 inClosure Size:38-400Color:naturalMaterial:polyethyleneStyle:funnelClosure Style:screw threadCountry of Origin:Made in the USA
Catalog No. NC3451535 Supplier California Pacific Lab & Consulting Supplier No. EF438400
4" ECO Funnel® with 38-400 cap adapter for 38mm plastic jugsSize:4 inClosure Size:38-400Color:naturalMaterial:polyethyleneStyle:funnelClosure Style:screw threadCountry of Origin:Made in the USA