Learn More
Roboz Surgical 4IN FORCEPTS 1.2MM
SDP
Supplier:  Roboz Surgical RS5210
Encompass
Bonaccolto Forceps; Longitudinal Serrations, Cross Serrated At Tip; 1.2mm Tip Width; 4 in. Length
Catalog No. NC9901249