Learn More
California Pacific Lab & Consulting 4IN FUNNEL SAFETY HAZARDOUS
SDP
Catalog No. 5059012
Encompass_Preferred
Eco funnel, 4 in. dia. w/38-430 screw cap. Made of HDPE and has an easy use lid, gasket, positive latch, filter, and an 11 in. stem to effectively reduce chemical evaporation of hazardous solvents in the lab, 250mL
Catalog No. 50-590-12 Supplier California Pacific Lab & Consulting Supplier No. EF438
Eco funnel, 4 in. dia. w/38-430 screw cap. Made of HDPE and has an easy use lid, gasket, positive latch, filter, and an 11 in. stem to effectively reduce chemical evaporation of hazardous solvents in the lab, 250mL