Learn More
Sklar Instruments Scissors, Iris Scissors, Sklar 4 in., Blunt/Blunt, Straight, 2 Probe Points, Stainless Steel, EA.
SDP
Catalog No. 501093948 Shop All Sklar Instruments Products
Encompass_Preferred
Scissors, Iris Scissors, Sklar, 4 in., Blunt/Blunt, Straight, 2 Probe Points, Stainless Steel, EA. Originally designed for ophthalmic procedures, Iris Scissors have become a multipurpose instrument used in various surgical environments. These fine, sharp scissors are ideal for detailed dissection of fine tissue and fine suture removal. In addition to ophthalmic procedures, Iris Scissors are now often used in emergency rooms, and in procedures involving fine tissue such as OB/GYN and dermatology. This product is straight with smooth, blunt/blunt tips and two probe points. It has a length of 4 inches.
Catalog No. 50-109-3948 Supplier Sklar Instruments Supplier No. 642046
Scissors, Iris Scissors, Sklar, 4 in., Blunt/Blunt, Straight, 2 Probe Points, Stainless Steel, EA. Originally designed for ophthalmic procedures, Iris Scissors have become a multipurpose instrument used in various surgical environments. These fine, sharp scissors are ideal for detailed dissection of fine tissue and fine suture removal. In addition to ophthalmic procedures, Iris Scissors are now often used in emergency rooms, and in procedures involving fine tissue such as OB/GYN and dermatology. This product is straight with smooth, blunt/blunt tips and two probe points. It has a length of 4 inches.