Learn More
THORLABS INC Ø1/2" Optical Post, SS, 8-32 Setscrew, 1/4"-20 Tap, L = 4"
Catalog No. NC9868022
Encompass
Ø1/2" Optical Post, SS, 8-32 Setscrew, 1/4"-20 Tap, L = 4"
Catalog No. NC9868022 Supplier THORLABS INC Supplier No. TR4
Ø1/2" Optical Post, SS, 8-32 Setscrew, 1/4"-20 Tap, L = 4"