Learn More
Ace Glass, Inc. Flange clamp, 100mm (4in), release
SDP
Catalog No. 509557344
Encompass
100mm (4) stainless steel quick release flange clamp without support rod, for use with two-piece pressure reaction flasks and for heads with flanges.
Catalog No. 50-955-7344 Supplier Ace Glass, Inc. Supplier No. 651725
100mm (4) stainless steel quick release flange clamp without support rod, for use with two-piece pressure reaction flasks and for heads with flanges.