Learn More
LICOR 4-Inch Soft Roller (1 roller)
Catalog No. NC9304708
Encompass_Preferred
The roller is used to eliminate air bubbles from membranes on the glass surface of the Odyssey M or Odyssey DLx Imager. It can also be used when placing membranes or gels on the Odyssey XF imaging tray (P/N 926-40020).
Catalog No. NC9304708 Supplier LICOR Supplier No. 92671000
The roller is used to eliminate air bubbles from membranes on the glass surface of the Odyssey M or Odyssey DLx Imager. It can also be used when placing membranes or gels on the Odyssey XF imaging tray (P/N 926-40020).