Learn More
Cole-Parmer Wika 332.54 4 4" High-Precision Test Gauge, 0 to 160 psi
Supplier:  Cole-Parmer 6843221
Encompass_Preferred
Stainless Steel wetted parts - .025% Accuracy WIKA inspector’s test gauges are convenient for field calibrations. They have an accuracy of ±0.25% which meets ASME B40.1 Grade 3A. The mirrored band on the dial and the knife-edge pointer make it easy to take accurate readings from the gauge. These test gauges are supplied standard with a zippered, nylon carrying pouch.
Catalog No. 50-992-580