Learn More
Uline Gusseted Reclosable Bags - 4 x 6 x 2,1000
Catalog No. NC2051494
Encompass_Preferred
Gusseted bottom lets product stand upright,Double-track zipper is ultra easy to use,100 virgin polyethylene,Meets FDA and USDA specifications,LDPE Low Density Polyethylene,Sulfur free,Amine and Silicone free,Latex free,BHT free Butylated Hydroxy Toluene,Chlorine free,Recyclable,Side seal,Shipped with zippers closed Open bags not available,Can be put in freezer but cannot be quickflash frozen,Not treated for printing by end-user,Not recommended for dry ice,Not microwaveable,Cannot be used for baking cooking or boiling,Not leakproof waterproof or airtight,Acid free and archival safe
Catalog No. NC2051494 Supplier Uline Supplier No. S10709
Gusseted bottom lets product stand upright,Double-track zipper is ultra easy to use,100 virgin polyethylene,Meets FDA and USDA specifications,LDPE Low Density Polyethylene,Sulfur free,Amine and Silicone free,Latex free,BHT free Butylated Hydroxy Toluene,Chlorine free,Recyclable,Side seal,Shipped with zippers closed Open bags not available,Can be put in freezer but cannot be quickflash frozen,Not treated for printing by end-user,Not recommended for dry ice,Not microwaveable,Cannot be used for baking cooking or boiling,Not leakproof waterproof or airtight,Acid free and archival safe