Learn More
Chemglass Life Sciences 4000mL Erlenmeyer Flask, 38-430 GPI Screw Thread
SDP
Catalog No. CG154308
Encompass_Preferred
4000mL Erlenmeyer Flask, 38-430 GPI Screw Thread. Erlenmeyer flask with screw thread finish. Supplied complete with white rubber lined black phenolic cap. Long skirted caps can be loosened without falling off while shaking flask.
Catalog No. CG154308 Supplier Chemglass Life Sciences Supplier No. CG154308
4000mL Erlenmeyer Flask, 38-430 GPI Screw Thread. Erlenmeyer flask with screw thread finish. Supplied complete with white rubber lined black phenolic cap. Long skirted caps can be loosened without falling off while shaking flask.