Learn More
REPLIGEN CORPORATION Repligen ACBT-4TC 4L Reservoir, Non-Irradiated, Polypropylene, 4 Ports
SDP
Catalog No. NC1782585
Encompass
The ACBT-4TC 4L Flat Bottom Reservoir from Repligen Corporation delivers reliable fluid management for scientific applications. Constructed from durable polypropylene, it features versatile tubing connections and supports a maximum working volume of 4000 mL with a flow rate of 8 L/min. This non-irradiated reservoir ensures excellent chemical compatibility and is ideal for various laboratory processes, setting it apart from other options in the market.
Catalog No. NC1782585 Supplier REPLIGEN CORPORATION Supplier No. ACBT4TC
The ACBT-4TC 4L Flat Bottom Reservoir from Repligen Corporation delivers reliable fluid management for scientific applications. Constructed from durable polypropylene, it features versatile tubing connections and supports a maximum working volume of 4000 mL with a flow rate of 8 L/min. This non-irradiated reservoir ensures excellent chemical compatibility and is ideal for various laboratory processes, setting it apart from other options in the market.