Learn More
Quality Environmental Containers 128oz/4000mL Natural Round Wide Mouth (Level 2) | 4/case
SDP
Catalog No. NC1835417
Encompass_Preferred
HDPE Round Wide-mouth containers are assembled, with polyethylene foam-lined polypropylene cap. Level 2 products are pre-cleaned to meet or exceed EPA requirements, and a Certificate of Quality Environmental Compliance (C-QEC) is issued. The C-QEC has a production lot number and is included in every case. Each case is labeled with the production lot number, custody-sealed and shipped with labels and custody seals.
Catalog No. NC1835417 Supplier Quality Environmental Containers Supplier No. 22130128
HDPE Round Wide-mouth containers are assembled, with polyethylene foam-lined polypropylene cap. Level 2 products are pre-cleaned to meet or exceed EPA requirements, and a Certificate of Quality Environmental Compliance (C-QEC) is issued. The C-QEC has a production lot number and is included in every case. Each case is labeled with the production lot number, custody-sealed and shipped with labels and custody seals.