Learn More
Diversified Biotech Inc 4L ICEPAN,BLU,5/CS
Catalog No. NC1605895
Encompass
IcePan, Cooling, Triple Density Expanded Polystyrene Ice lasts 30% longer, sweatless, durable, tolerates dry ice, liquid nitrogen, slurry 4L GLACIERBRAND ICE PAN, BLUE
Catalog No. NC1605895 Supplier Diversified Biotech Inc Supplier No. IPAN2000CS
IcePan, Cooling, Triple Density Expanded Polystyrene Ice lasts 30% longer, sweatless, durable, tolerates dry ice, liquid nitrogen, slurry 4L GLACIERBRAND ICE PAN, BLUE