Learn More
Ace Glass, Inc. Beaker, 4L, jacketed, Ace-Safe hose connections for 1/2intubing, 232mm inside height, 150mm inside diameter
SDP
Catalog No. NC0144511
Encompass_Preferred
4L jacketed beaker with upper and lower Ace-Safe hose connections for 1/2 in. tubing. Dimensions: 232mm inside height X 150mm inside diameter. Does not have pouring spout.
Catalog No. NC0144511 Supplier Ace Glass, Inc. Supplier No. 5340130
4L jacketed beaker with upper and lower Ace-Safe hose connections for 1/2 in. tubing. Dimensions: 232mm inside height X 150mm inside diameter. Does not have pouring spout.