Learn More
Ace Glass, Inc. 4000mL separatory funnel, pear shaped, complete with #15removable PTFE drip tip and 6mm bore PTFE stopcock plug, 12632-38 size 38 PTFE stopper not included
SDP
Catalog No. 50979212
Encompass_Preferred
4000ml pear-shaped, separatory funnel complete with #15 removable PTFE drip tip and 6mm bore PTFE stopcock plug. Drip tip is detachable, difficult to break and easy to clean.
Catalog No. 50-979-212 Supplier Ace Glass, Inc. Supplier No. 722366
4000ml pear-shaped, separatory funnel complete with #15 removable PTFE drip tip and 6mm bore PTFE stopcock plug. Drip tip is detachable, difficult to break and easy to clean.