Learn More
Andwin Scientific 4LB TERG-A-ZYME POWDER
SDP
Catalog No. NC9567330
Encompass_Preferred
The Tergazyme Enzyme-Action Powder Detergent is ideal as a cleaning agent in reverse osmosis and ultra-filtration equipment or for presoaking hard-to-remove stains. It eliminates proteinaceous soils, tissue, blood, and body fluids from several surfaces like glass, metal, stainless steel, porcelain, ceramic, plastic, rubber, and fiberglass, without leaving any residue that may intervene any later procedure. This is packaged in a box and is sold in a quantity of 1.8 kilograms or 4 pounds.
This item is not returnable. View return policy
Catalog No. NC9567330 Supplier Andwin Scientific Supplier No. 21837118
The Tergazyme Enzyme-Action Powder Detergent is ideal as a cleaning agent in reverse osmosis and ultra-filtration equipment or for presoaking hard-to-remove stains. It eliminates proteinaceous soils, tissue, blood, and body fluids from several surfaces like glass, metal, stainless steel, porcelain, ceramic, plastic, rubber, and fiberglass, without leaving any residue that may intervene any later procedure. This is packaged in a box and is sold in a quantity of 1.8 kilograms or 4 pounds.