Learn More
ATCC 4MBr-5; Lung; Rhesus Monkey (Macaca mulatta)
Catalog No. 502383227
Encompass
4MBr-5 is an epithelial cell line that was isolated in 1978 from the normal bronchial tissue of a rhesus monkey. This cell line was deposited by JL Caputo.
Catalog No. 50-238-3227 Supplier ATCC Supplier No. CCL208
4MBr-5 is an epithelial cell line that was isolated in 1978 from the normal bronchial tissue of a rhesus monkey. This cell line was deposited by JL Caputo.