Learn More
Avanti Polar Lipids 1,2-diphytanoyl-sn-glycero-3-phosphocholine, 207131-40-6, MFCD00674308, 25 mg
Catalog No. NC1075660
Encompass
Empirical Formula (Hill Notation): C48H96NO8P, Molecular Weight: 846.25, Purity >99%. Lipids containing diphytanoyl fatty acid chains have been used to produce stable planar lipid membranes (see References). Diphytanoyl phosphatidylcholine does not exhibit a detectable gel to liquid crystalline phase transition from -120°C to +120°C.
Catalog No. NC1075660 Supplier Avanti Polar Lipids Supplier No. 850356C25MG
Empirical Formula (Hill Notation): C48H96NO8P, Molecular Weight: 846.25, Purity >99%. Lipids containing diphytanoyl fatty acid chains have been used to produce stable planar lipid membranes (see References). Diphytanoyl phosphatidylcholine does not exhibit a detectable gel to liquid crystalline phase transition from -120°C to +120°C.