Learn More
Cayman Chemical 4methoxy5HydroxycnthIn6 10mg
SDP
Catalog No. 502942144
Encompass_Preferred
An alkaloid with anti-inflammatory activity; reduces LPS-induced TNF-α release from, and NO production in, RAW 264.7 macrophages at 10, 30, and 100 µM; reduces paw swelling in a rat model of CFA-induced arthritis at 9 and 27 mg/kg; reduces colonic inflammatory cytokine levels and maintains mucosal barrier function in a mouse model of DSS-induced colitis from 25-100 mg/kg
Catalog No. 50-294-2144 Supplier Cayman Chemical Supplier No. 4023410 MG
An alkaloid with anti-inflammatory activity; reduces LPS-induced TNF-α release from, and NO production in, RAW 264.7 macrophages at 10, 30, and 100 µM; reduces paw swelling in a rat model of CFA-induced arthritis at 9 and 27 mg/kg; reduces colonic inflammatory cytokine levels and maintains mucosal barrier function in a mouse model of DSS-induced colitis from 25-100 mg/kg