Learn More
Cayman Chemical 4-methoxy-α-Pyrrolidinopentiophenone (hydrochloride), Purity: ≥98%, CAS: 5537-19-9, 10 mg
SDP
Catalog No. 502953824
Encompass_Preferred
Shares structural features with the stimulant α-PVP, whose mechanism of action is unknown but is believed to act similarly to the designer drug MDPV
Catalog No. 50-295-3824 Supplier Cayman Chemical Supplier No. 1409710 MG
Shares structural features with the stimulant α-PVP, whose mechanism of action is unknown but is believed to act similarly to the designer drug MDPV