Learn More
Cayman Chemical 4methoxyaPyrroldInopentIo 50mg
SDP
Catalog No. 502955487
Encompass_Preferred
Shares structural features with the stimulant α-PVP, whose mechanism of action is unknown but is believed to act similarly to the designer drug MDPV
Catalog No. 50-295-5487 Supplier Cayman Chemical Supplier No. 1409750 MG
Shares structural features with the stimulant α-PVP, whose mechanism of action is unknown but is believed to act similarly to the designer drug MDPV