Learn More
Cayman Chemical 4methoxyN NDImthylcathIno 10mg
SDP
Catalog No. 502951691
Encompass_Preferred
An analog of methedrone, differing by the presence of a second aminomethyl group; intended for forensic and research applications
Catalog No. 50-295-1691 Supplier Cayman Chemical Supplier No. 1166610 MG
An analog of methedrone, differing by the presence of a second aminomethyl group; intended for forensic and research applications