Learn More
Cayman Chemical 4methoxyN NDImthylcathIno 50mg
SDP
Catalog No. 502951803
Encompass_Preferred
An analog of methedrone, differing by the presence of a second aminomethyl group; intended for forensic and research applications
Catalog No. 50-295-1803 Supplier Cayman Chemical Supplier No. 1166650 MG
An analog of methedrone, differing by the presence of a second aminomethyl group; intended for forensic and research applications