Learn More
Cayman Chemical 4methyl Fentnylhydrochlo 100ug
SDP
Catalog No. 502912331
Encompass_Preferred
An analytical reference standard; structurally similar to known opioids; provided as a DEA exempt preparation; intended for research and forensic applications
Catalog No. 50-291-2331 Supplier Cayman Chemical Supplier No. 36374100 UG
An analytical reference standard; structurally similar to known opioids; provided as a DEA exempt preparation; intended for research and forensic applications