Learn More
Cayman Chemical 4Methyl7 2nItrophenoxy2 10mg
SDP
Catalog No. 502938952
Encompass_Preferred
A fluorescent coumarin derivative; ex/em = 323/445 nm, respectively
Catalog No. 50-293-8952 Supplier Cayman Chemical Supplier No. 2847310 MG
A fluorescent coumarin derivative; ex/em = 323/445 nm, respectively