Learn More
Cayman Chemical 4-Methyl-7-(4-nitro-2-(trifluoromethyl)phenoxy)-2H-chromen-2-one, Purity: ≥98%, CAS: 433254-36-5, 5 mg
SDP
Catalog No. 502940112
Encompass_Preferred
A turn-on fluorescent probe for H2S; excited at 348 nm in DMSO and fluorescence intensity increases in the presence of HS-; selective for HS- over cysteine, AcO-, F-, H2PO4-, Cl-, Br-, and I- and is not cytotoxic up to a concentration of 150 µg/ml
Catalog No. 50-294-0112 Supplier Cayman Chemical Supplier No. 284715 MG
A turn-on fluorescent probe for H2S; excited at 348 nm in DMSO and fluorescence intensity increases in the presence of HS-; selective for HS- over cysteine, AcO-, F-, H2PO4-, Cl-, Br-, and I- and is not cytotoxic up to a concentration of 150 µg/ml