Learn More
Cayman Chemical 4-Methyl-7-(4-nitrophenoxy)-2H-chromen-2-one, Purity: ≥98%, CAS: 1644277-00-8, 10 mg
SDP
Catalog No. 502938942
Encompass_Preferred
A fluorescent coumarin derivative; ex/em = 323/445 nm, respectively
Catalog No. 50-293-8942 Supplier Cayman Chemical Supplier No. 2847410 MG
A fluorescent coumarin derivative; ex/em = 323/445 nm, respectively