Learn More
Cayman Chemical 4MethylethcathInon metabolI
SDP
Catalog No. 502970691
Encompass_Preferred
A metabolite of 4-MEC that features conversion of the β-keto group to β-hydroxy and is an enantiomeric mixture of the R,S and S,R orientations at carbons one and two, as in ephedrine; intended for research and forensic applications
Catalog No. 50-297-0691 Supplier Cayman Chemical Supplier No. 14758500 UG
A metabolite of 4-MEC that features conversion of the β-keto group to β-hydroxy and is an enantiomeric mixture of the R,S and S,R orientations at carbons one and two, as in ephedrine; intended for research and forensic applications