Learn More
Cayman Chemical 4-Methylumbelliferyl 2-Acetamido-2-deoxy-α-D-galactopyranoside, Purity: ≥98%, CAS: 124223-99-0, 10 mg
SDP
Catalog No. 502995651
Encompass_Preferred
A fluorogenic substrate for α- and β-galactopyranosaminidase; is cleaved by α- and β-galactopyranosaminidase to release the fluorescent moiety 4-MU
Catalog No. 50-299-5651 Supplier Cayman Chemical Supplier No. 2309910 MG
A fluorogenic substrate for α- and β-galactopyranosaminidase; is cleaved by α- and β-galactopyranosaminidase to release the fluorescent moiety 4-MU