Learn More
Cayman Chemical 4-Methylumbelliferyl Caprylate, Purity: ≥98%, CAS: 20671-66-3, 10 g
SDP
Catalog No. 502932700
Encompass_Preferred
A fluorogenic substrate for C8 esterase; is cleaved by C8 esterase to release the fluorescent moiety 4-MU; ex = 320 and 360 nm at low (1.97-6.72) and high (7.12-10.3) pH, respectively; em = 445 to 455 nm, increasing as pH decreases
Catalog No. 50-293-2700 Supplier Cayman Chemical Supplier No. 2680210 G
A fluorogenic substrate for C8 esterase; is cleaved by C8 esterase to release the fluorescent moiety 4-MU; ex = 320 and 360 nm at low (1.97-6.72) and high (7.12-10.3) pH, respectively; em = 445 to 455 nm, increasing as pH decreases