Learn More
Cayman Chemical 4-Methylumbelliferyl β-D-Galactopyranoside-6-sulfate (sodium salt), Purity: ≥98%, CAS: 206443-06-3, 10 mg
SDP
Catalog No. 502983289
Encompass_Preferred
A fluorogenic substrate used to quantify GALNS activity; is cleaved by GALNS to release the fluorescent moiety 4-MU; ex = 320 and 360 nm at low (1.97-6.72) and high (7.12-10.3) pH, respectively; em = 445-455 nm, increasing as pH decreases; used to detect Morquio disease type A in an assay using a very small blood volume
Catalog No. 50-298-3289 Supplier Cayman Chemical Supplier No. 2090710 MG
A fluorogenic substrate used to quantify GALNS activity; is cleaved by GALNS to release the fluorescent moiety 4-MU; ex = 320 and 360 nm at low (1.97-6.72) and high (7.12-10.3) pH, respectively; em = 445-455 nm, increasing as pH decreases; used to detect Morquio disease type A in an assay using a very small blood volume