Learn More
Cayman Chemical 4-Methylumbelliferyl Oleate, Purity: ≥98%, CAS: 18323-58-5, 100 mg
SDP
Catalog No. 502983741
Encompass_Preferred
A fluorogenic substrate for acid and alkaline lipases
Catalog No. 50-298-3741 Supplier Cayman Chemical Supplier No. 19590100 MG
A fluorogenic substrate for acid and alkaline lipases